Orders LinkContactUsLinkComponent
Combustion boats
Combustion boats
103×20×12 mm
Catalog # TSLA73382
Combustion boats
Catalog # TSLA73382
Combustion boats
103×20×12 mm
Price
147,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Combustion boats, quartz, glazed, with or without handle.
  • Very good chemical resistance
  • Very good temperature resistance
Specifications
  • Seq:
    02
  • W×D×H:
    103×20×12 mm