Order Entry
Export
Orders LinkContactUsLinkComponent
VWR® Dip Slides

VWR® Dip Slides

Plate count agar and VRBGA with neutralisers, for Staphylococcus aureus and Enterobacteriaceae, 100
Catalog # 535174R
Supplier:   VWR Chemicals
VWR® Dip Slides
Catalog # 535174R
Supplier:  VWR Chemicals
CAS Number:  
VWR® Dip Slides
VWR® Dip Slides

Plate count agar and VRBGA with neutralisers, for Staphylococcus aureus and Enterobacteriaceae, 100

CAS Number:  
Price
188,00 €
Log in for availability
1 * 100 items
Product Details & Documents

Dip slides for microbiological monitoring of surfaces and in liquids. Each slide has media on both sides, with a choice of the same medium, or two different media on the two sides of the slide. This allows sampling of two surfaces with every slide. Testing with dip slides is a safe, reliable, economical and fast indicator of the presence of microorganisms. Dip slides, with a push cap, have a larger contact area.

  • Supplied sterile
  • Sampling takes only a few seconds
  • Results are easily read after just 24 to 48 hours
Specifications

Specifications

  • Pk:
    100
  • Description:
    Plate count agar and VRBGA with neutralisers, for Staphylococcus aureus and Enterobacteriaceae
  • Cat. No.:
    535174R
  • Type:
    Push cap
  • Paddle size:
    80×28 mm|80×28
  • Dimensions:
    30 mm tube, 110 mm high
  • Tray W×L:
    50×22 mm
  • Contact area:
    10 cm²|10

Specifications