VWR® Dip Slides
Plate count agar MacConkey agar with neutralisers, for total count, Salmonella, Shigella and Coliform, 10
Dip slides for microbiological monitoring of surfaces and in liquids. Each slide has media on both sides, with a choice of the same medium, or two different media on the two sides of the slide. This allows sampling of two surfaces with every slide. Testing with dip slides is a safe, reliable, economical and fast indicator of the presence of microorganisms. Dip slides, with a push cap, have a larger contact area.
- Supplied sterile
- Sampling takes only a few seconds
- Results are easily read after just 24 to 48 hours
Pack insert TDS Nutrient Agar with TTC and Malt Extract Agar Large Dip Slides (535102B) TDS Nutrient Agar with TTC and Malt Extract Agar, Pack of 10 TDS Plate Count Agar with Neutraliser Black D TDS Plate Count Agar with Neutraliser Black DI
- Pk:10
- Description:Plate count agar MacConkey agar with neutralisers, for total count, Salmonella, Shigella and Coliform
- Cat. No.:535152L
- Type:Push cap
- Paddle size:80×28 mm|80×28
- Dimensions:30 mm tube, 110 mm high
- Tray W×L:50×22 mm
- Contact area:10 cm²|10



