VWR® Dip Slides
TSA with TTC and Rose Bengal agar, for total count, yeasts and moulds, 10
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Dip slides for microbiological monitoring of surfaces and in liquids. Each slide has media on both sides, with a choice of the same medium, or two different media on the two sides of the slide. This allows sampling of two surfaces with every slide. Testing with dip slides is a safe, reliable, economical and fast indicator of the presence of microorganisms. Dip slides, with a push cap, have a larger contact area.
- Supplied sterile
- Sampling takes only a few seconds
- Results are easily read after just 24 to 48 hours
Documents
Certificates
Search by Lot NumberPack insert TDS Nutrient Agar with TTC and Malt Extract Agar Large Dip Slides (535102B) TDS Nutrient Agar with TTC and Malt Extract Agar, Pack of 10 TDS Plate Count Agar with Neutraliser Black D TDS Plate Count Agar with Neutraliser Black DI
Datasheet
More
SDS_EN_CH_VWRC535092Q SDS_EN_CH_VWRC535092Q.pdf SDS_EN_CH_VWRC535104D SDS_EN_CH_VWRC535202E SDS_EN_CH_VWRC535202E.pdf
SDS
More
Spec VWRC535314D.pdf spec_VWRC535124H spec_VWRC535134J spec_VWRC535142J spec_VWRC535202E
Product specification
Specifications
Specifications
- Pk:10
- Description:TSA with TTC and Rose Bengal agar, for total count, yeasts and moulds
- Cat. No.:535112D
- Type:Push cap
- Paddle size:80×28 mm|80×28
- Dimensions:30 mm tube, 110 mm high
- Tray W×L:50×22 mm
- Contact area:10 cm²|10



