Order Entry
Export
Orders LinkContactUsLinkComponent
Accessories for flat cleanroom mops

Accessories for flat cleanroom mops

SlimLine™ Stainless steel mop frame, 360° swivel, quick disconnect, 9" flat head mops; SSU series handles, 9×4" (22.9×10 cm) mm
Catalog # 129-0536
   Waste Reducing
Supplier:   MICRONOVA
Accessories for flat cleanroom mops
Catalog # 129-0536
Supplier:  MICRONOVA
   Waste Reducing
Accessories for flat cleanroom mops

SlimLine™ Stainless steel mop frame, 360° swivel, quick disconnect, 9" flat head mops; SSU series handles, 9×4" (22.9×10 cm) mm

Price
379,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Description:
    SlimLine™ Stainless steel mop frame, 360° swivel, quick disconnect, 9" flat head mops; SSU series handles
  • L×W:
    9×4" (22.9×10 cm) mm
  • Seq:
    21
  • Cat. No.:
    129-0536
  • Environmentally Preferable:
    Waste Reducing