Order Entry
Export
Orders LinkContactUsLinkComponent
Test tubes with rim, medium wall

Test tubes with rim, medium wall

150 mm, 18 mm
Catalog # 212-7407
Supplier:   SciLabware
Test tubes with rim, medium wall
Catalog # 212-7407
Supplier:  SciLabware
Test tubes with rim, medium wall
Test tubes with rim, medium wall

150 mm, 18 mm

Price
121,00 €
Log in for availability
1 * 100 items
Product Details & Documents

PYREX®, borosilicate glass.

  • Superior performance in comparison to neutral borosilicate and soda-lime glass
  • Excellent resistance to chemicals and high temperature

High quality test tubes designed and constructed for optimum performance.

Certifications: ISO 4142

Specifications
  • Ext.Ø:
    18 mm
  • Length:
    150 mm
  • Seq:
    08