Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,3-Diethylurea ≥97%

Glass bottle for solids, 25 g
Catalog # L02079.14
1,3-Diethylurea ≥97%
Catalog # L02079.14
CAS Number:  623-76-7
1,3-Diethylurea ≥97%
1,3-Diethylurea ≥97%

Glass bottle for solids, 25 g

CAS Number:  623-76-7
Price
152,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 97% CAS No.: 623-76-7 Molecular Formula: C5H12N2O Formula Weight: 116.16 Melting Point: 109-113° Beilstein Reference: 1744741

Synonyms:  N,N'-Diethylurea
  • CAS Number:623-76-7
  • Formula:C₅H₁₂N₂O
  • MDL Number:MFCD00009028
  • Molecular Weight:116.16 g/mol
  • Melting Pt:109…113 °C
  • Storage Temperature:Ambient
  • EINECS:210-811-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications