Order Entry
Export
Orders LinkContactUsLinkComponent
Ethyl-(S)-(-)-lactate ≥99%

Ethyl-(S)-(-)-lactate ≥99%

Glass bottle for solids, 500 g
Catalog # A10900.36
Corrosive
Flammable
Irritant
Ethyl-(S)-(-)-lactate ≥99%
Catalog # A10900.36
CAS Number:  687-47-8
Ethyl-(S)-(-)-lactate ≥99%
Ethyl-(S)-(-)-lactate ≥99%

Glass bottle for solids, 500 g

CAS Number:  687-47-8
Price
39,40 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 99% CAS No.: 687-47-8 Molecular Formula: C5H10O3 Formula Weight: 118.13 Flash Point: 46°(114°F) Physical Form: Liquid Density: 1.036 Refractive Index: 1.4130 Appearance: Colourless Melting Point: -25° Boiling Point: 152-155° Beilstein Reference: 1720839 Merck Reference: 14,3817 Optical Rotation: -11° (neat) UN No.: 1192 MDL No.: MFCD00065359

Synonyms:  Ethyl (S)-(-)-2-hydroxypropionate, Ethyl L(-)-lactate, (S)-Ethyl 2-hydroxypropanoate, (S)-(-)-Ethyl lactate
  • CAS Number:687-47-8
  • Formula:C₅H₁₀O₃
  • MDL Number:MFCD00004518
  • Molecular Weight:118.13 g/mol
  • Merck Index:14,3817
  • Boiling Pt:152…155 °C
  • Melting Pt:–25 °C
  • Density:1.036
  • Flash Pt:46 °C (114 °F)
  • Storage Temperature:Ambient
  • EINECS:211-694-1
  • UN:1192
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications