Order Entry
Export
Orders LinkContactUsLinkComponent
1,9-Diaminononane 98%

1,9-Diaminononane 98%

Glass bottle for solids, 25 g
Catalog # L02043.14
Corrosive
1,9-Diaminononane 98%
Catalog # L02043.14
CAS Number:  646-24-2
1,9-Diaminononane 98%
1,9-Diaminononane 98%

Glass bottle for solids, 25 g

CAS Number:  646-24-2
Price
709,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98% CAS No.: 646-24-2 Molecular Formula: H2N(CH2)9NH2 Formula Weight: 158.29 Flash Point: 110°(230°F) Melting Point: 36-40° Boiling Point: 258-259° Beilstein Reference: 1737533 UN No.: 3259 MDL No.: MFCD00008251

Synonyms:  1,9-Nonanediamine, Nonamethylendiamine
  • CAS Number:646-24-2
  • Formula:C₉H₂₂N₂
  • MDL Number:MFCD00008251
  • Molecular Weight:158.29 g/mol
  • Boiling Pt:258…259 °C
  • Melting Pt:36…40 °C
  • Flash Pt:>110 °C (230 °F)
  • Storage Temperature:Ambient
  • EINECS:211-470-3
  • UN:3259
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications