Orders LinkContactUsLinkComponent
1,9-Diaminononane 98%
Glass bottle for solids, 5 g
  L02043.06
1,9-Diaminononane 98%
  L02043.06
 :  646-24-2
1,9-Diaminononane 98%
1,9-Diaminononane 98%
Glass bottle for solids, 5 g
 :  646-24-2
181,00 €
1 * 5 g
Purity: 98% CAS No.: 646-24-2 Molecular Formula: H2N(CH2)9NH2 Formula Weight: 158.29 Flash Point: 110°(230°F) Melting Point: 36-40° Boiling Point: 258-259° Beilstein Reference: 1737533 UN No.: 3259 MDL No.: MFCD00008251
 :  1,9-Nonanediamine, Nonamethylendiamine
  •  :646-24-2
  •  :C₉H₂₂N₂
  •  :MFCD00008251
  •  :158.29 g/mol
  •  :258…259 °C
  •  :36…40 °C
  •  :>110 °C (230 °F)
  •  :Ambient
  •  :211-470-3
  •  :3259
  •  :8,III
  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g