Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,3-Dihydroxynaphthalene ≥98%

Glass bottle for solids, 5 g
Catalog # A17739.06
Irritant
1,3-Dihydroxynaphthalene ≥98%
Catalog # A17739.06
CAS Number:  132-86-5
1,3-Dihydroxynaphthalene ≥98%
1,3-Dihydroxynaphthalene ≥98%

Glass bottle for solids, 5 g

CAS Number:  132-86-5
Price
183,00 €
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 98% CAS No.: 132-86-5 Molecular Formula: C10H8O2 Formula Weight: 160.17 Melting Point: 123-126° Beilstein Reference: 2044002 Merck Reference: 14,6396 MDL No.: MFCD00003965

Synonyms:  Naphthalene-1,3-diol, Naphthalene-1,3-diol
  • CAS Number:132-86-5
  • Formula:C₁₀H₈O₂
  • MDL Number:MFCD00003965
  • Molecular Weight:160.17 g/mol
  • Merck Index:14,6396
  • Boiling Pt:386 °C (1013 hPa)
  • Melting Pt:123…126 °C
  • Density:1.12 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:205-079-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Cat. No.:
    A17739.06

Specifications