Orders LinkContactUsLinkComponent
Joint clips for conical joints, PTFE/steel spring, Quickfit®
Joint clips for conical joints, PTFE/steel spring, Quickfit®
White, 10/19
Catalog # 201-3025
Supplier:   SciLabware
Joint clips for conical joints, PTFE/steel spring, Quickfit®
Catalog # 201-3025
Supplier:  SciLabware
Joint clips for conical joints, PTFE/steel spring, Quickfit®
Joint clips for conical joints, PTFE/steel spring, Quickfit®
White, 10/19
Price
44,10 €
Log in for availability
1 * 5 items
Product Details & Documents
Clips for retaining glassware having Quickfit® conical joints. Manufactured from PTFE with encapsulated steel spring to provide a firm grip.
  • Excellent chemical resistance
  • Can be used at temperatures up to 250 °C
  • Clips are numbered with joint size for easy identification
Specifications
  • For NS:
    10/19
  • Colour:
    White
  • Seq:
    01