Orders LinkContactUsLinkComponent
Stirrer blades, PTFE, Quickfit®
Stirrer blades, PTFE, Quickfit®
Stirrer blade, with 24/29 socket, to fit ST6/7, 76 mm
Catalog # 441-0360
Supplier:   SciLabware
Stirrer blades, PTFE, Quickfit®
Catalog # 441-0360
Supplier:  SciLabware
Stirrer blades, PTFE, Quickfit®
Stirrer blades, PTFE, Quickfit®
Stirrer blade, with 24/29 socket, to fit ST6/7, 76 mm
Price
54,50 €
Log in for availability
1 * 5 items
Product Details & Documents
Stirrer blades made of PTFE, and they can be pivoted on the glass stirrer shaft to allow passage through small apertures.
  • Easily detachable for cleaning and replacement purposes
  • Resistance to chemical attack
Suitable for sweeping close to the vessel walls as they will not cause scratching.
Specifications
  • Type:
    Stirrer blade, with 24/29 socket, to fit ST6/7
  • Width:
    76 mm
  • Seq:
    02