Powder funnels, Quickfit®
19/26 NS, 105 mm
Catalog # 201-8295
Supplier: SciLabware
Jump to:
Product Details & Documents
PYREX®, borosilicate glass, stem with cone.
- Very good chemical resistance
- High temperature resistance
Designed for transfer of powders into flasks and vessels.
Specifications
- Cone size:19/26 NS
- Funnel O-Ø:105 mm
- Seq:02