Order Entry
Export
Orders LinkContactUsLinkComponent
(2-Methoxyethoxy)methyl chloride ≥94%

(2-Methoxyethoxy)methyl chloride ≥94%

Glass bottle for solids, 25 g
Catalog # L01050.14
Health
Flammable
Irritant
(2-Methoxyethoxy)methyl chloride ≥94%
Catalog # L01050.14
CAS Number:  3970-21-6
(2-Methoxyethoxy)methyl chloride ≥94%
(2-Methoxyethoxy)methyl chloride ≥94%

Glass bottle for solids, 25 g

CAS Number:  3970-21-6
Price
153,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 94% CAS No.: 3970-21-6 Molecular Formula: C4H9ClO2 Formula Weight: 124.57 Storage Temperature: -30°C to -10°C Flash Point: 54°(129°F) Refractive Index: 1.4270 Boiling Point: 50-52°/13mm Beilstein Reference: 1900576 UN No.: 1992 MDL No.: MFCD00000888

Synonyms:  1-Chloromethoxy-2-methoxyethane, 1-Chloro-2,5-dioxahexane, MEM chloride
  • CAS Number:3970-21-6
  • Formula:ClCH₂OCH₂CH₂OCH₃
  • MDL Number:MFCD00000888
  • Molecular Weight:124.57 g/mol
  • Boiling Pt:50…52 °C (13 mmHg)
  • Density:1.094
  • Flash Pt:54 °C (129 °F)
  • EINECS:223-589-8
  • UN:1992
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications