Soxhlet extractors, Quickfit®
Socket 100; cone 34/35; thimble size 95×200, 2000 ml
201-8372
Borosilicate glass, large volume.
Flanged extractors which require Quickfit® or NS jointed flasks and condensers.
- Extraction cap.:2000 ml
- Description:Socket 100; cone 34/35; thimble size 95×200