Orders LinkContactUsLinkComponent
Specimen bag re-sealable, BiTran®
Specimen re-sealable bags, 180×200 mm
Catalog # 720-0019
Supplier:   CELLPATH
Specimen bag re-sealable, BiTran®
Catalog # 720-0019
Supplier:  CELLPATH
Specimen bag re-sealable, BiTran®
Specimen bag re-sealable, BiTran®
Specimen re-sealable bags, 180×200 mm
Price
432,00 €
Log in for availability
1 * 250 items
Product Details & Documents
The BiTran bag system is designed for transporting wet tissue from one place to another and for the short to long–term storage of tissue.
The bags are manufactured from a special barrier film with a unique double zip closure. The barrier film is impermeable to formalin vapour, therefore no hazardous vapour pollutes the laboratory or any other area once the bag is sealed. The double zip seal provides a leak proof closure, which can be easily opened and closed as many times as required.
  • Leak proof
  • Well–proven long–term storage
  • Space saving
  • High visibility barrier film
The BiTranStor Box is available for the long term storage of tissue in BiTran Bags. The boxes are portable and stackable and provides a cost–effective solution for storing BiTranbags.
Specifications
  • Description:
    Specimen re-sealable bags
  • Dimensions:
    180×200 mm