2-Phenylpropionic acid 98%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
2-Phenylpropionic acid 98%
Catalog # L00709.14
Supplier: Thermo Fisher Scientific
CAS Number: 492-37-5
Jump to:
Product Details & Documents
Purity: 98% CAS No.: 492-37-5 Molecular Formula: C9H10O2 Formula Weight: 150.18 Flash Point: 148°(298°F) Refractive Index: 1.5230 Melting Point: -5° Boiling Point: 264-266° Beilstein Reference: 1863558 MDL No.: MFCD00002650
Synonyms: (±)-2-Phenylpropionic acid, (±)-Hydratropic acid, DL-2-Phenylpropionic acid, (±)-2-Phenylpropanoic acid
- CAS Number:492-37-5
- Formula:C₉H₁₀O₂
- MDL Number:MFCD00002650
- Molecular Weight:150.18 g/mol
- Boiling Pt:264…266 °C
- Melting Pt:–5 °C
- Density:1.100
- Flash Pt:148 °C (298 °F)
- Storage Temperature:Ambient
- EINECS:207-752-0
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



