Order Entry
Export
Orders LinkContactUsLinkComponent
2-Phenylpropionic acid 98%

2-Phenylpropionic acid 98%

Glass bottle for solids, 5 g
Catalog # L00709.06
Irritant
2-Phenylpropionic acid 98%
Catalog # L00709.06
CAS Number:  492-37-5
2-Phenylpropionic acid 98%
2-Phenylpropionic acid 98%

Glass bottle for solids, 5 g

CAS Number:  492-37-5
Price
34,50 €
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 98% CAS No.: 492-37-5 Molecular Formula: C9H10O2 Formula Weight: 150.18 Flash Point: 148°(298°F) Refractive Index: 1.5230 Melting Point: -5° Boiling Point: 264-266° Beilstein Reference: 1863558 MDL No.: MFCD00002650

Synonyms:  (±)-2-Phenylpropionic acid, (±)-Hydratropic acid, DL-2-Phenylpropionic acid, (±)-2-Phenylpropanoic acid
  • CAS Number:492-37-5
  • Formula:C₉H₁₀O₂
  • MDL Number:MFCD00002650
  • Molecular Weight:150.18 g/mol
  • Boiling Pt:264…266 °C
  • Melting Pt:–5 °C
  • Density:1.100
  • Flash Pt:148 °C (298 °F)
  • Storage Temperature:Ambient
  • EINECS:207-752-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications