Order Entry
Export
Orders LinkContactUsLinkComponent
 

Aspirating pipettes, Falcon®

5 ml, 348 mm
Catalog # 612-2458
Supplier:   Corning
Aspirating pipettes, Falcon®
Catalog # 612-2458
Supplier:  Corning
Aspirating pipettes, Falcon®
Aspirating pipettes, Falcon®

5 ml, 348 mm

Price
65,18 €
Log in for availability
1 * 200 items
Product Details & Documents

PS pipettes, non plugged, non graduated, disposable, sterile.

  • Safer alternative to glass Pasteur pipettes
  • Non pyrogenic
  • Individually packed to prevent contamination

 : Individually packed in a thermo-formed paper/plastic wrap.

Specifications
  • Capacity:
    5 ml
  • Length:
    348 mm