Pressure Bulbs
Atomiser bulb with pressure valve, red-brown, size 5, PVC, 102
Catalog # DENE4200005
Supplier: DEUTSCH NEUMANN
Jump to:
Product Details & Documents
Natural rubber, chloroprene or PVC.
Used for filling automatic burettes, Pellet pattern (ball set with net) or atomisers (bulb).
Specifications
- Capacity:102
- Description:Atomiser bulb with pressure valve, red-brown, size 5, PVC
- Seq:3