Order Entry
Export
Orders LinkContactUsLinkComponent
Cell culture inserts, Falcon®

Cell culture inserts, Falcon®

Inserts for 12-well plates, sterile, Transparent PET membrane, 1,0 μm
Catalog # 734-0041
Supplier:   Corning
Cell culture inserts, Falcon®
Catalog # 734-0041
Supplier:  Corning
Cell culture inserts, Falcon®
Cell culture inserts, Falcon®

Inserts for 12-well plates, sterile, Transparent PET membrane, 1,0 μm

Price
334,22 €
Log in for availability
1 * 48 items
Product Details & Documents

Track-etched PET membranes have a smooth surface and defined cylindrical pores that traverse the membrane.

  • Low protein binding PET membrane
  • Sterilized by gamma irradiation
  • A wide variety of configurations including 6, 12 and 24 well
  • A broad selection of membrane pore sizes, 0,4, 1,0, 3,0 and 8,0 µm diameter
  • Packed in individual blister packs, 48 inserts/case
  • Non-Tissue Culture-treated insert housings prevent promiscuous growth of cells on the insert walls
  • Innovative hanging design facilitates pipetting and allows for co-culture
Specifications
  • Membrane:
    Transparent PET membrane
  • Pore size:
    1,0 μm
  • Description:
    Inserts for 12-well plates, sterile
  • Seq:
    08