Order Entry
Export
Orders LinkContactUsLinkComponent
 

Volumetric flasks, class A, white markings

5 ml, 10/13, ± 0,025 ml
  612-1856
Volumetric flasks, class A, white markings
  612-1856
Volumetric flasks, class A, white markings
Volumetric flasks, class A, white markings

5 ml, 10/13, ± 0,025 ml

 
120,00 €
Log in for availability
1 * 5 items
 

Borosilicate glass, amber, with PP NS-stopper.

  • Easy to read white markings
  • Very good chemical resistance
  • Very good temperature resistance

Amber coloured to protect light sensitive substances.

 : Acc. to ISO and ASTM or USP standards

Specifications
  • NS:
    10/13
  • Capacity:
    5 ml
  • Seq:
    01
  • Tolerance:
    ± 0,025 ml