Burettes, removable stopcock
PTFE stopcock
Catalog # 612-0376
Supplier: SciLabware
Jump to:
Product Details & Documents
Soda lime glass, class B, removable PTFE stopcock.
- Flared top for safer filling
- Blue enamel graduations
BS 846
Certifications: BS 846
Documents
Specifications
- Description:PTFE stopcock
- Seq:06