Order Entry
Export
Orders LinkContactUsLinkComponent
1,9-Nonanedithiol ≥97%

1,9-Nonanedithiol ≥97%

Glass bottle for solids, 1 g
Catalog # L07172.03
Toxic
Irritant
1,9-Nonanedithiol ≥97%
Catalog # L07172.03
CAS Number:  3489-28-9
1,9-Nonanedithiol ≥97%
1,9-Nonanedithiol ≥97%

Glass bottle for solids, 1 g

CAS Number:  3489-28-9
Price
58,00 €
Log in for availability
1 * 1 g
Product Details & Documents

Purity: 97% CAS No.: 3489-28-9 Molecular Formula: HS(CH2)9SH Formula Weight: 192.39 Flash Point: 100°(212°F) Refractive Index: 1.4999 Boiling Point: 351-352° Beilstein Reference: 1737568 MDL No.: MFCD00004914

Synonyms:  1,9-Dimercaptononane
  • CAS Number:3489-28-9
  • Formula:C₉H₂₀S₂
  • MDL Number:MFCD00004914
  • Molecular Weight:192.39 g/mol
  • Boiling Pt:351…352 °C
  • Melting Pt:–17.5 °C
  • Density:0.953
  • Flash Pt:>100 °C (212 °F)
  • Storage Temperature:Ambient
  • EINECS:222-482-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications