7-Methylindole ≥98%
Glass bottle for solids, 1 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 98% CAS No.: 933-67-5 Molecular Formula: C9H9N Formula Weight: 131.18 Melting Point: 83-84° Boiling Point: 266° MDL No.: MFCD00005684
- CAS Number:933-67-5
- Formula:C₉H₉N
- MDL Number:MFCD00005684
- Molecular Weight:131.18 g/mol
- Boiling Pt:266 °C
- Melting Pt:83…84 °C
- Density:1.0202 g/cm³ (99.6 °C)
- :Density: 1.0202 g/cm³ (99.6 °C)
- Storage Temperature:Ambient
- EINECS:213-270-1
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



