Order Entry
Export
Orders LinkContactUsLinkComponent
3-(Hydroxymethyl)indole ≥97%

3-(Hydroxymethyl)indole ≥97%

Glass bottle for solids, 5 g
Catalog # L15762.06
3-(Hydroxymethyl)indole ≥97%
Catalog # L15762.06
CAS Number:  700-06-1
3-(Hydroxymethyl)indole ≥97%
3-(Hydroxymethyl)indole ≥97%

Glass bottle for solids, 5 g

CAS Number:  700-06-1
Price
76,10 €
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 97% CAS No.: 700-06-1 Molecular Formula: C9H9NO Formula Weight: 147.18 Melting Point: 96-99° Beilstein Reference: 121323

Synonyms:  3-Indolmethanol, 3-Indolcarbinol, (1H-Indol-3-yl)methanol, Indole-3-methanol, (Indole-3-yl)methanol, 3-Indolemethanol, Indole-3-carbinol
  • CAS Number:700-06-1
  • Formula:C₉H₉NO
  • MDL Number:MFCD00005632
  • Molecular Weight:147.18 g/mol
  • Melting Pt:96…99 °C
  • Storage Temperature:Ambient
  • EINECS:211-836-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications