Order Entry
Export
Orders LinkContactUsLinkComponent
2,3-Dimethylfuran ≥99%

2,3-Dimethylfuran ≥99%

Glass bottle for solids, 5 g
Catalog # L15705.06
Flammable
Irritant
2,3-Dimethylfuran ≥99%
Catalog # L15705.06
CAS Number:  14920-89-9
2,3-Dimethylfuran ≥99%
2,3-Dimethylfuran ≥99%

Glass bottle for solids, 5 g

CAS Number:  14920-89-9
Price
147,00 €
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 99% CAS No.: 14920-89-9 Molecular Formula: C6H8O Formula Weight: 96.13 Flash Point: -1°(30°F) Refractive Index: 1.4430 Boiling Point: 42°/15mm Beilstein Reference: 104572 UN No.: 1992 MDL No.: MFCD00153893

  • CAS Number:14920-89-9
  • Formula:C₆H₈O
  • MDL Number:MFCD00153893
  • Molecular Weight:96.13 g/mol
  • Boiling Pt:42 °C (115 mmHg)
  • Density:0.910
  • Flash Pt:–1 °C (30 °F)
  • Storage Temperature:Ambient
  • UN:1992
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications