Order Entry
Export
Orders LinkContactUsLinkComponent
 

Polysorbate 60

Glass bottle, 500 mL
Catalog # L15031.AP
Polysorbate 60
Catalog # L15031.AP
CAS Number:  9005-67-8
Polysorbate 60
Polysorbate 60

Glass bottle, 500 mL

CAS Number:  9005-67-8
Price
45,70 €
Log in for availability
1 * 500 mL
Product Details & Documents

CAS No.: 9005-67-8 Molecular Formula: C64H126O26 Flash Point: 150°(302°F) Merck Reference: 14,7582 MDL No.: MFCD01780110

Synonyms:  Sorbitan monostearate, ethoxylated, Polyoxyethylene sorbitan monostearate
  • CAS Number:9005-67-8
  • Formula:C₆₄H₁₂₆O₂₆
  • MDL Number:MFCD01780110
  • Merck Index:14,7582
  • Density:1.080
  • Flash Pt:>150 °C (302 °F)
  • Storage Temperature:Ambient
  • EINECS:500-020-4
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications