Poly(methylhydrosiloxane)
50 g
Supplier: Thermo Fisher Scientific
Poly(methylhydrosiloxane)
Catalog # L14561.18
Supplier: Thermo Fisher Scientific
CAS Number: 63148-57-2
Jump to:
Product Details & Documents
Poly(methylhydrosiloxane) is used in the reduction of esters to alcohols, catalyzed by a combination of titanocene dichloride and either n-BuLi or EtMgBr. It is a safer alternative to triethoxysilane, B22063, for reduction of phosphine oxides to phosphines, catalyzed by Ti(O-i-Pr)4. It is also used in greener amine synthesis by reductive amination as an alternative to borohydrides.
CAS No.: 63148-57-2 Molecular Formula: (CH3)3SiO[(CH3)HSiO]nSi(CH3) Formula Weight: ca 1900 Flash Point: 204°(399°F) Refractive Index: 1.3980
Synonyms: Methyl hydrogen polysiloxane, PMHS
- CAS Number:63148-57-2
- Formula:(CH₃)₃SiO[(CH₃)HSiO]nSi(CH₃)
- MDL Number:MFCD00084478
- Boiling Pt:∼140 °C (0.003 hPa)
- Melting Pt:–55 °C
- Density:1.006
- Flash Pt:204 °C (399 °F)
- Storage Temperature:Ambient
Specifications
Specifications
- Pk:50 g



