Iron(III) bromide, anhydrous 98+%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
Iron(III) bromide, anhydrous 98+%
Catalog # 31125.06
Supplier: Thermo Fisher Scientific
CAS Number: 10031-26-2
Jump to:
Product Details & Documents
Purity: 98+% CAS No.: 10031-26-2 Molecular Formula: FeBr3 Formula Weight: 295.57 Physical Form: Crystalline Sensitivity: Light Sensitive and Hygroscopic Appearance: Dark red Odour: Odourless Melting Point: dec. Merck Reference: 14,4018 MDL No.: MFCD00016082
Synonyms: Ferric bromide, Iron tribromide
- CAS Number:10031-26-2
- Formula:Br₃Fe
- MDL Number:MFCD00016082
- Molecular Weight:295.56 g/mol
- Merck Index:14,4018
- Melting Pt:Dec.
- Storage Temperature:Ambient
- EINECS:233-089-1
- UN:3260
- ADR:8,III
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



