Orders LinkContactUsLinkComponent
Sodium peroxide 95%
Glass bottle for solids, 250 g
  L13306.30
Sodium peroxide 95%
  L13306.30
 :  1313-60-6
Sodium peroxide 95%
Sodium peroxide 95%
Glass bottle for solids, 250 g
 :  1313-60-6
174,00 €
1 * 250 g
Purity: 95% CAS No.: 1313-60-6 Molecular Formula: Na2O2 Formula Weight: 77.98 Melting Point: 460° dec. Merck Reference: 14,8655 MDL No.: MFCD00003497
 :  di-Sodium peroxide
  •  :1313-60-6
  •  :Na₂O₂
  •  :MFCD00003497
  •  :77.98 g/mol
  •  :14,8655
  •  :460 °C (dec.)
  •  :2.805
  •  :Ambient
  •  :215-209-4
  •  :1504
  •  :5.1,I
  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g