Order Entry
Export
Orders LinkContactUsLinkComponent
Ammonium hexafluorosilicate ≥98%

Ammonium hexafluorosilicate ≥98%

Glass bottle for solids, 250 g
Catalog # 69100.30
Toxic
Ammonium hexafluorosilicate ≥98%
Catalog # 69100.30
CAS Number:  16919-19-0
Ammonium hexafluorosilicate ≥98%
Ammonium hexafluorosilicate ≥98%

Glass bottle for solids, 250 g

CAS Number:  16919-19-0
Price
49,40 €
Log in for availability
1 * 250 g
Product Details & Documents

Purity: 98% CAS No.: 16919-19-0 Molecular Formula: (NH4)2SiF6 Formula Weight: 178.14 Physical Form: Powder Melting Point: dec. UN No.: 2854 MDL No.: MFCD00010887

Etching glass, in soldering flux

Synonyms:  Ammonium silicofluoride Ammonium hexafluorosilicate, Ammonium fluorosilicate, Ammomium fluosilicate
  • CAS Number:16919-19-0
  • Formula:H₈F₆N₂Si
  • MDL Number:MFCD00010887
  • Molecular Weight:178.15 g/mol
  • Merck Index:14,527
  • Melting Pt:Dec.
  • Density:2.01 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:240-968-3
  • UN:2854
  • ADR:6.1,III
  •  :Density: 2.01 g/cm³ (20 °C)
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications