Methanol ≥99%
Glass bottle, 500 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 67-56-1 Molecular Formula: CH3OH Formula Weight: 32.04 Flash Point: 11°(52°F) Refractive Index: 1.3290 Odour: Mild Melting Point: -98° Boiling Point: 64.7° Beilstein Reference: 1098229 Merck Reference: 14,5957 UN No.: 1230 MDL No.: MFCD00004595
Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
- CAS Number:67-56-1
- Formula:CH₃OH
- MDL Number:MFCD00004595
- Molecular Weight:32.04 g/mol
- Merck Index:14,5957
- Boiling Pt:64.7 °C
- Melting Pt:–98 °C
- Density:0.791
- Flash Pt:11 °C (52 °F)
- Storage Temperature:Ambient
- EINECS:200-659-6
- UN:1230
- ADR:3,II
- REACH:01-2119433307-44
Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:500 mL



