Order Entry
Export
Orders LinkContactUsLinkComponent
Fluorescein ≥90%

Fluorescein ≥90%

Glass bottle for solids, 500 g
Catalog # L13251.36
Irritant
Fluorescein ≥90%
Catalog # L13251.36
CAS Number:  2321-07-5
Fluorescein ≥90%
Fluorescein ≥90%

Glass bottle for solids, 500 g

CAS Number:  2321-07-5
Price
132,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 90+% CAS No.: 2321-07-5 Molecular Formula: C20H12O5 Formula Weight: 332.32 Melting Point: 320° Beilstein Reference: 94324 Merck Reference: 14,4159 MDL No.: MFCD00005050

Synonyms:  3',6'-Dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 2-(6-Hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoic acid
  • CAS Number:2321-07-5
  • Formula:C₂₀H₁₂O₅
  • MDL Number:MFCD00005050
  • Molecular Weight:332.31 g/mol
  • Merck Index:14,4159
  • Melting Pt:320 °C
  • Storage Temperature:Ambient
  • EINECS:219-031-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications