Order Entry
Export
Orders LinkContactUsLinkComponent
1,4-Dioxane ≥99% for HPLC

1,4-Dioxane ≥99% for HPLC

Glass bottle, 1 L
Catalog # 43167.K2
Flammable
Health
Irritant
1,4-Dioxane ≥99% for HPLC
Catalog # 43167.K2
CAS Number:  123-91-1
1,4-Dioxane ≥99% for HPLC
1,4-Dioxane ≥99% for HPLC

Glass bottle, 1 L

CAS Number:  123-91-1
Price
135,00 €
Log in for availability
1 * 1 L
Product Details & Documents

Purity: 99% min CAS No.: 123-91-1 Molecular Formula: C4H8O2 Formula Weight: 88.11 Flash Point: 12°(54°F) Physical Form: Liquid Density: 1.034 Refractive Index: 1.4220 Sensitivity: Hygroscopic Odour: Ether-like Melting Point: 11.8° Boiling Point: 100-102° Beilstein Reference: 102551 Merck Reference: 14,3300 UN No.: 1165 MDL No.: MFCD00006571

Synonyms:  Glycolethylether, 1,4-Dioxacyclohexane, Dioxane, p-Dioxane, 1,4-Diethylene dioxide
  • CAS Number:123-91-1
  • Formula:C₄H₈O₂
  • MDL Number:MFCD00006571
  • Molecular Weight:88.11 g/mol
  • Merck Index:14,3300
  • Boiling Pt:100…102 °C
  • Melting Pt:11.8 °C
  • Density:1.034
  • Flash Pt:12 °C (54 °F)
  • Storage Temperature:Ambient
  • UN:1165
  • ADR:3,II
  •  :Packaged under argon in resealable ChemSeal™ bottles.
  • EINECS:204-661-8
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications