Order Entry
Export
Orders LinkContactUsLinkComponent
Dichloromethane ≥99% stabilised

Dichloromethane ≥99% stabilised

Glass bottle, 500 mL
Catalog # L13089.AP
Health
Irritant
Dichloromethane ≥99% stabilised
Catalog # L13089.AP
CAS Number:  75-09-2
Dichloromethane ≥99% stabilised
Dichloromethane ≥99% stabilised

Glass bottle, 500 mL

CAS Number:  75-09-2
Price
29,50 €
Log in for availability
1 * 500 mL
Product Details & Documents

Stabilized with ca. 50 ppm amylene

Purity: 99+% CAS No.: 75-09-2 Molecular Formula: CH2Cl2 Formula Weight: 84.93 Refractive Index: 1.4244 Odour: Sweet Boiling Point: 39-40° Beilstein Reference: 1730800 Merck Reference: 14,6063 UN No.: 1593 MDL No.: MFCD00000881

Synonyms:  Methylene chloride, DCM
  • CAS Number:75-09-2
  • Formula:CH₂Cl₂
  • MDL Number:MFCD00000881
  • Molecular Weight:84.93 g/mol
  • Merck Index:14,6063
  • Boiling Pt:39…40 °C
  • Melting Pt:–95 °C
  • Density:1.325
  • Storage Temperature:Ambient
  • EINECS:200-838-9
  • ADR:6.1,III
  •  :Stabilised with 2-methyl-2-butene (iso-amylene) 50 ppm
  • UN:1593
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications