Orders LinkContactUsLinkComponent
1,2-Ethanedithiol ≥98%
Glass bottle for solids, 25 g
  L12865.14
1,2-Ethanedithiol ≥98%
  L12865.14
 :  540-63-6
1,2-Ethanedithiol ≥98%
1,2-Ethanedithiol ≥98%
Glass bottle for solids, 25 g
 :  540-63-6
30,80 €
1 * 25 g
Purity: 98+% CAS No.: 540-63-6 Molecular Formula: C2H6S2 Formula Weight: 94.20 Flash Point: 45°(113°F) Refractive Index: 1.5580 Odour: Stench Melting Point: -41° Boiling Point: 144-146° Beilstein Reference: 505946 Merck Reference: 14,3725 UN No.: 3071 MDL No.: MFCD00004892
 :  1.2-Ethanedithiol, 1,2-Dimercaptoethane
  •  :540-63-6
  •  :C₂H₆S₂
  •  :MFCD00004892
  •  :94.2 g/mol
  •  :14,3725
  •  :144…146 °C
  •  :–41 °C
  •  :1.123
  •  :45 °C (113 °F)
  •  :Ambient
  •  :208-752-3
  •  :3071
  •  :6.1,II
  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g