Order Entry
Export
Orders LinkContactUsLinkComponent
Bis-trispropane ≥98%

Bis-trispropane ≥98%

Glass bottle for solids, 25 g
Catalog # 43496.14
Bis-trispropane ≥98%
Catalog # 43496.14
CAS Number:  64431-96-5
Bis-trispropane ≥98%
Bis-trispropane ≥98%

Glass bottle for solids, 25 g

CAS Number:  64431-96-5
Price
67,10 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98+% CAS No.: 64431-96-5 Molecular Formula: CH2[CH2NHC(CH2OH)3]2 Formula Weight: 282.34 Physical Form: Powder Melting Point: 168-170° MDL No.: MFCD00004689

Synonyms:  1,3-Bis(tris(hydroxymethyl)methylamino)propane, 1,3-Bis[tris(hydroxymethyl)amino]propane, 2,2'-(Propane-1,3-diylbis(azanediyl))bis(2-(hydroxymethyl)propane-1,3-diol), BIS-TRIS propane, 2,2'-(Propane-1,3-diyldiimino)bis[2-(hydroxymethyl)propane-1,3-diol]
  • CAS Number:64431-96-5
  • Formula:C₁₁H₂₆N₂O₆
  • MDL Number:MFCD00004689
  • Molecular Weight:282.34 g/mol
  • Melting Pt:168…170 °C
  • Storage Temperature:Ambient
  • EINECS:264-899-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications