Order Entry
Export
Orders LinkContactUsLinkComponent
 

Bisacrylamide ≥99%

Glass bottle for solids, 25 g
Catalog # 43701.14
Irritant
Bisacrylamide ≥99%
Catalog # 43701.14
CAS Number:  110-26-9
Bisacrylamide ≥99%
Bisacrylamide ≥99%

Glass bottle for solids, 25 g

CAS Number:  110-26-9
Price
56,50 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 99+% CAS No.: 110-26-9 Molecular Formula: C7H10N2O2 Formula Weight: 154.17 Physical Form: Powder Density: 1.235 Appearance: White Melting Point: 300° Beilstein Reference: 1706297 MDL No.: MFCD00008625

Synonyms:  Bis (N,N'-methylenebisacrylamide), N,N-Methylenebisacrylamide, N,N'-Methylenediacrylamide, N,N’-Methylenebisacrylamide, N,N'-Methylenebisacrylamide, BIS
  • CAS Number:110-26-9
  • Formula:(H₂C=CHCONH)₂CH₂
  • MDL Number:MFCD00008625
  • Molecular Weight:154.17 g/mol
  • Boiling Pt:125 °C (3.33 hPa)
  • Melting Pt:>300 °C
  • Density:1.235
  • Flash Pt:138 °C
  • Storage Temperature:Ambient
  • EINECS:203-750-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications