Order Entry
Export
Orders LinkContactUsLinkComponent
 

Bis(pinacolato)diboron ≥98%

Glass bottle for solids, 5 g
Catalog # L16088.06
Irritant
Bis(pinacolato)diboron ≥98%
Catalog # L16088.06
CAS Number:  73183-34-3
Bis(pinacolato)diboron ≥98%
Bis(pinacolato)diboron ≥98%

Glass bottle for solids, 5 g

CAS Number:  73183-34-3
Price
147,00 €
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 98+% CAS No.: 73183-34-3 Molecular Formula: C12H24B2O4 Formula Weight: 253.94 Storage Temperature: 2°C to 8°C Melting Point: 133-139° Beilstein Reference: 7703552 MDL No.: MFCD00799570

Synonyms:  4,4,4',4',5,5,5',5'-Octamethyl-2,2'-bi(1,3,2-dioxaborolane), 4,4,5,5-tetramethyl-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3,2-dioxaborolane
  • CAS Number:73183-34-3
  • Formula:C₁₂H₂₄B₂O₄
  • MDL Number:MFCD00799570
  • Molecular Weight:253.94 g/mol
  • Melting Pt:133…139 °C
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications