Order Entry
Export
Orders LinkContactUsLinkComponent
1,3-Diphenylacetone ≥98%

1,3-Diphenylacetone ≥98%

Glass bottle for solids, 100 g
Catalog # L02830.22
1,3-Diphenylacetone ≥98%
Catalog # L02830.22
CAS Number:  102-04-5
1,3-Diphenylacetone ≥98%
1,3-Diphenylacetone ≥98%

Glass bottle for solids, 100 g

CAS Number:  102-04-5
Price
192,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 98+% CAS No.: 102-04-5 Molecular Formula: C15H14O Formula Weight: 210.28 Flash Point: 110°(230°F) Melting Point: 32-36° Boiling Point: 330° Beilstein Reference: 974767 MDL No.: MFCD00004795

Synonyms:  1,3-Diphenylpropan-2-one, 1,3-Diphenyl-2-propanone
  • CAS Number:102-04-5
  • Formula:(C₆H₅)CH₂C(O)CH₂(C₆H₅)
  • MDL Number:MFCD00004795
  • Molecular Weight:210.28 g/mol
  • Boiling Pt:330 °C
  • Melting Pt:32…36 °C
  • Density:1.04
  • Flash Pt:>110 °C (230 °F)
  • Storage Temperature:Ambient
  • EINECS:203-000-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications