Order Entry
Export
Orders LinkContactUsLinkComponent
2-Ethyl-1-butene ≥97%

2-Ethyl-1-butene ≥97%

Glass bottle for solids, 25 g
Catalog # L11553.14
Flammable
Irritant
2-Ethyl-1-butene ≥97%
Catalog # L11553.14
CAS Number:  760-21-4
2-Ethyl-1-butene ≥97%
2-Ethyl-1-butene ≥97%

Glass bottle for solids, 25 g

CAS Number:  760-21-4
Price
210,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 97% CAS No.: 760-21-4 Molecular Formula: C6H12 Formula Weight: 84.16 Flash Point: -26°(-14°F) Refractive Index: 1.3960 Melting Point: -132° Boiling Point: 63-64° Beilstein Reference: 1697102 UN No.: 3295 MDL No.: MFCD00009341

  • CAS Number:760-21-4
  • Formula:C₆H₁₂
  • MDL Number:MFCD00009341
  • Molecular Weight:84.16 g/mol
  • Boiling Pt:63…64 °C
  • Melting Pt:–132 °C
  • Density:0.689
  • Flash Pt:–26 °C (–14 °F)
  • Storage Temperature:Ambient
  • EINECS:212-078-5
  • UN:3295
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications