Order Entry
Export
Orders LinkContactUsLinkComponent
 

Dimetridazole ≥97%

Glass bottle for solids, 25 g
Catalog # L11303.14
Health
Dimetridazole ≥97%
Catalog # L11303.14
CAS Number:  551-92-8
Dimetridazole ≥97%
Dimetridazole ≥97%

Glass bottle for solids, 25 g

CAS Number:  551-92-8
Price
96,50 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 97% CAS No.: 551-92-8 Molecular Formula: C5H7N3O2 Formula Weight: 141.13 Melting Point: 136-138° Beilstein Reference: 130665 Merck Reference: 14,3265 MDL No.: MFCD00047046

Synonyms:  1,2-Dimethyl-5-nitroimidazole
  • CAS Number:551-92-8
  • Formula:C₅H₇N₃O₂
  • MDL Number:MFCD00047046
  • Molecular Weight:141.13 g/mol
  • Merck Index:14,3265
  • Melting Pt:136…138 °C
  • Storage Temperature:Ambient
  • EINECS:209-001-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications