Dimetridazole ≥97%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 97% CAS No.: 551-92-8 Molecular Formula: C5H7N3O2 Formula Weight: 141.13 Melting Point: 136-138° Beilstein Reference: 130665 Merck Reference: 14,3265 MDL No.: MFCD00047046
Synonyms: 1,2-Dimethyl-5-nitroimidazole
- CAS Number:551-92-8
- Formula:C₅H₇N₃O₂
- MDL Number:MFCD00047046
- Molecular Weight:141.13 g/mol
- Merck Index:14,3265
- Melting Pt:136…138 °C
- Storage Temperature:Ambient
- EINECS:209-001-2
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



