Order Entry
Export
Orders LinkContactUsLinkComponent
Ethyl acetate ≥99%

Ethyl acetate ≥99%

Glass bottle, 250 mL
Catalog # L10925.AK
Flammable
Irritant
   Safer Human and Environmental Health
Ethyl acetate ≥99%
Catalog # L10925.AK
   Safer Human and Environmental Health
CAS Number:  141-78-6
Ethyl acetate ≥99%
Ethyl acetate ≥99%

Glass bottle, 250 mL

CAS Number:  141-78-6
Price
27,00 €
Log in for availability
1 * 250 mL
Product Details & Documents

Purity: 99% CAS No.: 141-78-6 Molecular Formula: C4H8O2 Formula Weight: 88.11 Flash Point: -3°(26°F) Refractive Index: 1.3720 Odour: Sweet Melting Point: -84° Boiling Point: 76-77° Beilstein Reference: 506104 Merck Reference: 14,3757 UN No.: 1173 MDL No.: MFCD00009171

Synonyms:  Acetic acid ethyl ester
  • CAS Number:141-78-6
  • Formula:CH₃COOC₂H₅
  • MDL Number:MFCD00009171
  • Molecular Weight:88.11 g/mol
  • Merck Index:14,3757
  • Boiling Pt:76…77 °C
  • Melting Pt:–84 °C
  • Density:0.902
  • Flash Pt:–3 °C (26 °F)
  • Storage Temperature:Ambient
  • EINECS:205-500-4
  • UN:1173
  • ADR:3,II
  • REACH:01-2119475103-46
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Cat. No.:
    L10925.AK
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications