Order Entry
Export
Orders LinkContactUsLinkComponent
3,5-Dimethylbenzaldehyde ≥98%

3,5-Dimethylbenzaldehyde ≥98%

Glass bottle for solids, 1 g
Catalog # L09748.03
3,5-Dimethylbenzaldehyde ≥98%
Catalog # L09748.03
CAS Number:  5779-95-3
3,5-Dimethylbenzaldehyde ≥98%
3,5-Dimethylbenzaldehyde ≥98%

Glass bottle for solids, 1 g

CAS Number:  5779-95-3
Price
53,20 €
Log in for availability
1 * 1 g
Product Details & Documents

Refractive Index: 1.5375
Melting Point: 8-10°
Boiling Point: 220-222°
Beilstein Reference: 2038840
MDL No.: MFCD00082776

  • CAS Number:5779-95-3
  • Formula:C₉H₁₀O
  • MDL Number:MFCD00082777
  • Molecular Weight:134.18 g/mol
  • Boiling Pt:220…222 °C
  • Melting Pt:8…10 °C
  • Density:1.09 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:675-424-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications