Order Entry
Export
Orders LinkContactUsLinkComponent
D-(+)-Cellobiose octaacetate

D-(+)-Cellobiose octaacetate

Glass bottle for solids, 25 g
Catalog # L08780.14
D-(+)-Cellobiose octaacetate
Catalog # L08780.14
CAS Number:  5346-90-7
D-(+)-Cellobiose octaacetate
D-(+)-Cellobiose octaacetate

Glass bottle for solids, 25 g

CAS Number:  5346-90-7
Price
128,00 €
Log in for availability
1 * 25 g
Product Details & Documents

CAS No.: 5346-90-7 Molecular Formula: C28H38O19 Formula Weight: 678.59 Melting Point: 224-231° Beilstein Reference: 79280 Merck Reference: 14,1961 Optical Rotation: +40° (c=1 in chloroform) MDL No.: MFCD00009600

  • CAS Number:5346-90-7
  • Formula:C₂₈H₃₈O₁₉
  • MDL Number:MFCD00009600
  • Molecular Weight:678.59 g/mol
  • Merck Index:14,1961
  • Melting Pt:224…231 °C
  • Storage Temperature:Ambient
  • EINECS:226-304-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications