Linoleic acid 95%
Glass bottle for solids, 25 g
Purity: 95% CAS No.: 60-33-3 Molecular Formula: C18H32O2 Formula Weight: 280.45 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.4699 Odour: Weak, characteristic Melting Point: -12° Boiling Point: 230°/16mm Beilstein Reference: 1727101 Merck Reference: 14,5505 MDL No.: MFCD00064241
: (9Z,12Z)-Octadeca-9,12-dienoic acid, cis,cis-9,12-Octadecadienoic acid, 9Z,12Z-Octadecadienoic acid
- :60-33-3
- :C₁₈H₃₂O₂
- :MFCD00064241
- :280.45 g/mol
- :14,5505
- :230 °C (16 mmHg)
- :–12 °C
- :0.902
- :>110 °C (230 °F)
- :Refrigerator
- :200-470-9
- Pack type:Glass bottle for solids
- Pk:25 g