Order Entry
Export
Orders LinkContactUsLinkComponent
 

1-Naphthohydrazide ≥98%

25 g
Catalog # L07942.14
Irritant
1-Naphthohydrazide ≥98%
Catalog # L07942.14
CAS Number:  43038-45-5
1-Naphthohydrazide ≥98%
1-Naphthohydrazide ≥98%

25 g

CAS Number:  43038-45-5
Price
223,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98+% CAS No.: 43038-45-5 Molecular Formula: C11H10N2O Formula Weight: 186.21 Melting Point: 166-168° Beilstein Reference: 2966714 MDL No.: MFCD00016804

Synonyms:  1-Naphthalenecarbohydrazide
  • CAS Number:43038-45-5
  • Formula:C₁₁H₁₀N₂O
  • MDL Number:MFCD00016804
  • Molecular Weight:186.21 g/mol
  • Melting Pt:166…168 °C
  • Storage Temperature:Ambient
  • EINECS:256-054-2
Specifications

Specifications

  • Pk:
    25 g

Specifications